Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AZIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293755.1 | NP_001280684.1 | 8 - 9 | 1282 | 84 | |
XM_015452248.1 | XP_015307734.1 | 8 - 9 | 1240 | 84 | ||
XM_015452249.1 | XP_015307735.1 | 8 - 9 | 1254 | 84 | ||
XM_015452251.1 | XP_015307737.1 | 9 - 10 | 1321 | 84 | ||
XM_015452253.1 | XP_015307739.1 | 8 - 9 | 1254 | 84 | ||
XM_015452254.1 | XP_015307740.1 | 7 - 8 | 1077 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Probe spans exons |