Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283872.1 | NP_001270801.1 | 1 - 2 | 150 | 102 | |
GenBank mRNA | AB070178.1 | - | 1 - 2 | 150 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |