Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001283883.1 | NP_001270812.1 | 7 - 8 | 1202 | 79 | |
XM_015452072.1 | XP_015307558.1 | 6 - 7 | 1340 | 79 | ||
GenBank mRNA | AB071080.1 | - | 7 - 8 | 1202 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |