Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acvr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007397.2 | NP_031423.1 | 3 - 4 | 417 | 107 | |
GenBank mRNA | AK137432.1 | - | 2 - 3 | 108 | ||
BC106189.1 | - | 3 - 4 | 474 | 30240596 | 107 | |
M84120.1 | - | 3 - 4 | 415 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |