Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007439.2 | NP_031465.2 | 7 - 8 | 2163 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons |