Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007576.3 | NP_031602.3 | 2 - 3 | 353 | 65 | |
GenBank mRNA | AB094485.1 | - | 334 | 65 | ||
AB094486.1 | - | 2 - 3 | 269 | 65 | ||
AK136735.1 | - | 2 - 3 | 267 | 65 | ||
AK149468.1 | - | 2 - 3 | 360 | 65 | ||
AK149477.1 | - | 2 - 3 | 270 | 65 | ||
AK162345.1 | - | 2 - 3 | 267 | 65 | ||
BC012257.1 | - | 2 - 3 | 325 | 4163203 | 65 | |
M17122.1 | - | 2 - 3 | 367 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |