Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138730.1 | - | 2 - 3 | 145 | 132 | |
AK151547.1 | - | 2 - 3 | 313 | 132 | ||
AK156667.1 | - | 2 - 3 | 308 | 132 | ||
AK160328.1 | - | 2 - 3 | 308 | 132 | ||
AK171877.1 | - | 2 - 3 | 305 | 132 | ||
BC061255.1 | - | 2 - 3 | 312 | 30251263 | 132 | |
U59463.1 | - | 2 - 3 | 298 | 132 | ||
Y13089.1 | - | 2 - 3 | 290 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |