Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpina6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007618.2 | NP_031644.1 | 1 - 2 | 31 | 88 | |
GenBank mRNA | AK145842.1 | - | 1 - 2 | 63 | 88 | |
BC013632.1 | - | 1 - 2 | 50 | 4194164 | 88 | |
X70533.1 | - | 1 - 2 | 31 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |