Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007631.2 | NP_031657.1 | 2 - 3 | 642 | 86 | |
GenBank mRNA | AK005352.1 | - | 2 - 3 | 336 | 86 | |
AK017834.1 | - | 2 - 3 | 606 | 86 | ||
AK053348.1 | - | 2 - 3 | 550 | 86 | ||
AK149905.1 | - | 2 - 3 | 549 | 86 | ||
AK164997.1 | - | 2 - 3 | 587 | 86 | ||
AK170833.1 | - | 2 - 3 | 523 | 86 | ||
BC044841.1 | - | 2 - 3 | 545 | 3155470 | 86 | |
BC060172.1 | - | 2 - 3 | 537 | 86 | ||
EU600170.1 | - | 2 - 3 | 642 | 86 | ||
M64403.1 | - | 2 - 3 | 597 | 86 | ||
S78355.1 | - | 2 - 3 | 547 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |