Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007633.2 | NP_031659.2 | 4 - 5 | 370 | 63 | |
GenBank mRNA | AK089950.1 | - | 4 - 5 | 302 | 63 | |
AK163046.1 | - | 4 - 5 | 359 | 63 | ||
AK165658.1 | - | 4 - 5 | 360 | 63 | ||
BC062152.1 | - | 4 - 5 | 397 | 6510163 | 63 | |
BC084588.1 | - | 4 - 5 | 414 | 6506897 | 63 | |
BC106191.1 | - | 4 - 5 | 370 | 6336006 | 63 | |
BC138660.1 | - | 3 - 4 | 203 | 8861680 | 63 | |
BC138662.1 | - | 3 - 4 | 203 | 8861683 | 63 | |
X75888.1 | - | 3 - 4 | 210 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |