Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007635.4 | NP_031661.3 | 1 - 2 | 230 | 68 | |
GenBank mRNA | AF005885.1 | - | 1 - 2 | 208 | 68 | |
AK041941.1 | - | 1 - 2 | 548 | 68 | ||
AK170651.1 | - | 1 - 2 | 214 | 68 | ||
BC060180.1 | - | 1 - 2 | 101 | 68 | ||
BC088980.1 | - | 1 - 2 | 36 | 5686564 | 68 | |
BY065574.1 | - | 1 - 2 | 230 | 68 | ||
U95826.1 | - | 1 - 2 | 128 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |