Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007668.3 | NP_031694.1 | 2 - 3 | 170 | 54 | |
GenBank mRNA | AK051790.1 | - | 2 - 3 | 170 | 54 | |
BC052007.1 | - | 2 - 3 | 144 | 5706932 | 54 | |
D29678.1 | - | 2 - 3 | 179 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |