Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007711.3 | NP_031737.1 | 6 - 7 | 1316 | 120 | |
NM_173873.1 | NP_776298.1 | 7 - 8 | 1492 | 120 | ||
NM_173874.1 | NP_776299.1 | 7 - 8 | 1492 | 120 | ||
NM_173876.3 | NP_776301.1 | 6 - 7 | 1037 | 120 | ||
GenBank mRNA | AF029347.1 | - | 6 - 7 | 940 | 120 | |
AK141136.1 | - | 7 - 8 | 1430 | 120 | ||
AK147364.1 | - | 6 - 7 | 1472 | 120 | ||
AK147548.1 | - | 6 - 7 | 1474 | 120 | ||
AK147621.1 | - | 6 - 7 | 1474 | 120 | ||
AK149276.1 | - | 7 - 8 | 1031 | 120 | ||
AK150688.1 | - | 7 - 8 | 1421 | 120 | ||
AK159698.1 | - | 7 - 8 | 1466 | 120 | ||
AK161327.1 | - | 7 - 8 | 1420 | 120 | ||
AK162639.1 | - | 7 - 8 | 1433 | 120 | ||
AK169297.1 | - | 7 - 8 | 1413 | 120 | ||
AK169516.1 | - | 2 - 3 | 304 | 120 | ||
BC057133.1 | - | 6 - 7 | 970 | 6833732 | 120 | |
BC057855.1 | - | 6 - 7 | 1132 | 120 | ||
CF536658.1 | - | 3 - 4 | 463 | 120 | ||
X78874.1 | - | 6 - 7 | 1141 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |