Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252651.1 | NP_001239580.1 | 1 - 2 | 132 | 183 | |
NM_007782.3 | NP_031808.2 | 1 - 2 | 132 | 183 | ||
GenBank mRNA | AK089305.1 | - | 1 - 2 | 77 | 184 | |
M58288.1 | - | 1 - 2 | 55 | 183 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |