Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007807.5 | NP_031833.3 | 5 - 6 | 563 | 87 | |
GenBank mRNA | AK033570.1 | - | 5 - 6 | 507 | 87 | |
AK053920.1 | - | 5 - 6 | 507 | 87 | ||
AK152257.1 | - | 5 - 6 | 534 | 87 | ||
AK153154.1 | - | 5 - 6 | 534 | 87 | ||
AK153259.1 | - | 5 - 6 | 534 | 87 | ||
AK154126.1 | - | 5 - 6 | 563 | 87 | ||
AK171526.1 | - | 5 - 6 | 549 | 87 | ||
BC042838.1 | - | 5 - 6 | 530 | 87 | ||
BC071229.1 | - | 5 - 6 | 518 | 4979804 | 87 | |
FJ168469.1 | - | 5 - 6 | 552 | 87 | ||
U43384.1 | - | 5 - 6 | 489 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |