Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dll3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007866.2 | NP_031892.2 | 7 - 8 | 1704 | 118 | |
GenBank mRNA | AB013440.1 | - | 7 - 8 | 1691 | 118 | |
AK144282.1 | - | 7 - 8 | 1705 | 118 | ||
BC052002.1 | - | 7 - 8 | 1681 | 6404029 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |