Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eef2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267710.1 | NP_001254639.1 | 2 - 3 | 507 | 67 | |
NM_001267711.1 | NP_001254640.1 | 2 - 3 | 502 | 67 | ||
NM_007908.4 | NP_031934.1 | 2 - 3 | 591 | 67 | ||
GenBank mRNA | AK028524.1 | - | 2 - 3 | 568 | 67 | |
AK080915.1 | - | 2 - 3 | 67 | |||
AK136015.1 | - | 2 - 3 | 503 | 67 | ||
BC003433.1 | - | 2 - 3 | 539 | 2646503 | 67 | |
BC055361.1 | - | 2 - 3 | 502 | 30046537 | 67 | |
CJ052887.1 | - | 2 - 3 | 503 | 67 | ||
U93848.1 | - | 1 - 2 | 245 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |