Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007936.3 | NP_031962.2 | 14 - 15 | 2557 | 106 | |
GenBank mRNA | AK028509.1 | - | 14 - 15 | 2550 | 106 | |
AK147698.1 | - | 14 - 15 | 2553 | 106 | ||
BC004782.1 | - | 14 - 15 | 2472 | 3588285 | 106 | |
BC052164.1 | - | 14 - 15 | 2529 | 6512978 | 106 | |
S57168.1 | - | 14 - 15 | 2557 | 106 | ||
X65138.1 | - | 14 - 15 | 2557 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |