Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161373.1 | NP_001154845.1 | 21 - 22 | 6524 | 68 | |
NM_001161374.1 | NP_001154846.1 | 21 - 22 | 6524 | 68 | ||
NM_007977.2 | NP_032003.2 | 22 - 23 | 6734 | 68 | ||
GenBank mRNA | BC138347.1 | - | 21 - 22 | 6405 | 8861367 | 68 |
BC145245.1 | - | 21 - 22 | 6405 | 9053797 | 68 | |
L05573.1 | - | 22 - 23 | 6732 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |