Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007983.2 | NP_032009.2 | 7 - 8 | 1055 | 87 | |
GenBank mRNA | AK053752.1 | - | 7 - 8 | 990 | 87 | |
BC065098.1 | - | 7 - 8 | 1055 | 30362116 | 87 | |
U39643.1 | - | 7 - 8 | 1065 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |