Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008001.4 | NP_032027.2 | 9 - 10 | 2198 | 72 | |
GenBank mRNA | AK148151.1 | - | 9 - 10 | 2199 | 72 | |
BC004838.1 | - | 1 - 2 | 88 | 72 | ||
BC011462.1 | - | 9 - 10 | 2135 | 3674293 | 72 | |
U22325.1 | - | 9 - 10 | 1797 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |