Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700025035.mm.1 |
TC0700002790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025035.mm.1 |
NM_001110252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025035.mm.1 |
NM_001276269.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025035.mm.1 |
NM_008281.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025034.mm.1 |
TC0700002790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025034.mm.1 |
NM_001110252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025034.mm.1 |
NM_001276269.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025034.mm.1 |
NM_008281.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025032.mm.1 |
TC0700002790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025032.mm.1 |
NM_001110252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025032.mm.1 |
NM_001276269.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025032.mm.1 |
NM_008281.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025033.mm.1 |
TC0700002790.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700025033.mm.1 |
NM_001110252.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025033.mm.1 |
NM_001276269.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700025033.mm.1 |
NM_008281.4 |
Refseq |