Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008312.4 | NP_032338.3 | 2 - 3 | 622 | 115 | |
GenBank mRNA | AK034512.1 | - | 2 - 3 | 624 | 115 | |
AK043320.1 | - | 2 - 3 | 623 | 115 | ||
AK079648.1 | - | 2 - 3 | 624 | 115 | ||
AK086348.1 | - | 2 - 3 | 621 | 115 | ||
BC141085.1 | - | 2 - 3 | 201 | 9055697 | 115 | |
M63685.1 | - | 2 - 3 | 485 | 115 | ||
X72230.1 | - | 2 - 3 | 486 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |