Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il12rb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008353.2 | NP_032379.2 | 1 - 2 | 166 | 63 | |
GenBank mRNA | AK137577.1 | - | 1 - 2 | 167 | 63 | |
AK171937.1 | - | 1 - 2 | 143 | 63 | ||
BC057212.1 | - | 1 - 2 | 149 | 5350447 | 63 | |
U23922.1 | - | 1 - 2 | 121 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |