Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600022094.mm.1 |
TC0600002877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022094.mm.1 |
NM_008370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022094.mm.1 |
NM_008370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022096.mm.1 |
TC0600002877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022096.mm.1 |
NM_008370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022096.mm.1 |
NM_008370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022098.mm.1 |
TC0600002877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022098.mm.1 |
NM_008370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022098.mm.1 |
NM_008370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022093.mm.1 |
TC0600002877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022093.mm.1 |
NM_008370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022093.mm.1 |
NM_008370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022099.mm.1 |
TC0600002877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022099.mm.1 |
NM_008370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022099.mm.1 |
NM_008370.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022095.mm.1 |
TC0600002877.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600022095.mm.1 |
NM_008370.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600022095.mm.1 |
NM_008370.2 |
Refseq |