Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inhba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008380.1 | NP_032406.1 | 1 - 2 | 61 | 89 | |
GenBank mRNA | AK134991.1 | - | 3 - 4 | 512 | 89 | |
BC053527.1 | - | 1 - 2 | 53 | 6528822 | 90 | |
X69619.1 | - | 1 - 2 | 61 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |