Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008406.3 | NP_032432.2 | 8 - 9 | 943 | 77 | |
GenBank mRNA | AK149500.1 | - | 8 - 9 | 944 | 77 | |
BC013465.1 | - | 8 - 9 | 927 | 77 | ||
BC028814.1 | - | 8 - 9 | 943 | 5099284 | 77 | |
X70391.1 | - | 8 - 9 | 923 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |