Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008466.5 | NP_032492.1 | 14 - 15 | 1368 | 123 | |
GenBank mRNA | AF020772.1 | - | 14 - 15 | 1212 | 123 | |
AK049373.1 | - | 14 - 15 | 1239 | 123 | ||
AK161664.1 | - | 14 - 15 | 1298 | 123 | ||
BC026885.1 | - | 14 - 15 | 1262 | 3979788 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |