Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008467.4 | NP_032493.1 | 11 - 12 | 1194 | 79 | |
GenBank mRNA | AF020771.1 | - | 11 - 12 | 902 | 79 | |
AK147920.1 | - | 11 - 12 | 1203 | 79 | ||
BC026821.1 | - | 11 - 12 | 989 | 3672708 | 79 | |
BC052162.1 | - | 11 - 12 | 1195 | 6510795 | 79 | |
CT010281.1 | - | 11 - 12 | 902 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |