Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145802.1 | NP_001139274.1 | 2 - 3 | 429 | 89 | |
NM_001145803.1 | NP_001139275.1 | 2 - 3 | 429 | 89 | ||
NM_008547.2 | NP_032573.2 | 1 - 2 | 324 | 89 | ||
GenBank mRNA | BC050009.1 | - | 2 - 3 | 368 | 89 | |
BQ930470.1 | - | 2 - 3 | 344 | 89 | ||
X66983.1 | - | 1 - 2 | 318 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |