Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mertk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008587.1 | NP_032613.1 | 18 - 19 | 77 | ||
GenBank mRNA | AK029009.1 | - | 7 - 8 | 77 | ||
AK030158.1 | - | 10 - 11 | 77 | |||
AK079252.1 | - | 1 - 2 | 77 | |||
L11625.1 | - | 9 - 10 | 1057 | 77 | ||
U21301.1 | - | 18 - 19 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus 1700 Probe spans exons |