Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008710.3 | NP_032736.2 | 1257 | 65 | ||
GenBank mRNA | AK050478.1 | - | 1162 | 65 | ||
AK088044.1 | - | 1239 | 65 | |||
AK090115.1 | - | 1260 | 65 | |||
AK129006.1 | - | 1234 | 65 | |||
AK159694.1 | - | 1243 | 65 | |||
AK168736.1 | - | 1040 | 65 | |||
BC008518.1 | - | 1220 | 3601702 | 65 | ||
Z49204.1 | - | 1164 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |