Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008720.2 | NP_032746.2 | 13 - 14 | 2275 | 69 | |
GenBank mRNA | AF003348.1 | - | 13 - 14 | 2252 | 69 | |
AK154282.1 | - | 13 - 14 | 2195 | 69 | ||
AK155379.1 | - | 13 - 14 | 2195 | 69 | ||
BC052437.1 | - | 13 - 14 | 2275 | 6405214 | 69 | |
BC054539.1 | - | 13 - 14 | 2275 | 6405214 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |