Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008808.3 | NP_032834.1 | 1 - 2 | 248 | 73 | |
GenBank mRNA | AY324648.1 | - | 1 - 2 | 237 | 73 | |
BC003817.1 | - | 1 - 2 | 248 | 3495629 | 73 | |
M29464.1 | - | 1 - 2 | 128 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |