Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdgfrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146268.1 | NP_001139740.1 | 4 - 5 | 1056 | 73 | |
NM_008809.2 | NP_032835.2 | 4 - 5 | 1056 | 73 | ||
GenBank mRNA | AK134785.1 | - | 5 - 6 | 1078 | 73 | |
AK143247.1 | - | 4 - 5 | 1036 | 73 | ||
BC055311.1 | - | 4 - 5 | 1068 | 30060666 | 73 | |
X04367.1 | - | 4 - 5 | 765 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |