Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008830.2 | NP_032856.2 | 21 - 22 | 2870 | 107 | |
GenBank mRNA | BC150687.1 | - | 21 - 22 | 2740 | 9087598 | 107 |
J03398.1 | - | 21 - 22 | 2870 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |