Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008859.2 | NP_032885.1 | 2 - 3 | 207 | 78 | |
GenBank mRNA | AK037664.1 | - | 2 - 3 | 279 | 78 | |
BC138552.1 | - | 1 - 2 | 138 | 8861572 | 78 | |
BC145769.1 | - | 1 - 2 | 138 | 40131072 | 78 | |
D11091.1 | - | 2 - 3 | 207 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |