Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008927.3 | NP_032953.1 | 5 - 6 | 887 | 66 | |
GenBank mRNA | AB041567.1 | - | 5 - 6 | 867 | 66 | |
AK146233.1 | - | 5 - 6 | 877 | 66 | ||
AK146931.1 | - | 5 - 6 | 880 | 66 | ||
AK154793.1 | - | 5 - 6 | 914 | 66 | ||
AK165664.1 | - | 4 - 5 | 421 | 66 | ||
AK165894.1 | - | 5 - 6 | 888 | 66 | ||
AK167336.1 | - | 5 - 6 | 854 | 66 | ||
BC051137.1 | - | 5 - 6 | 706 | 66 | ||
BC054754.1 | - | 5 - 6 | 666 | 6411664 | 66 | |
L02526.1 | - | 5 - 6 | 614 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |