Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008939.2 | NP_032965.1 | 9 - 10 | 1949 | 65 | |
GenBank mRNA | AK132633.1 | - | 9 - 10 | 1738 | 65 | |
AK205149.1 | - | 2 - 3 | 187 | 65 | ||
BC031429.1 | - | 9 - 10 | 1851 | 3665834 | 65 | |
D89871.1 | - | 9 - 10 | 1947 | 65 | ||
Y13192.1 | - | 9 - 10 | 1726 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |