Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgds | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145834.1 | NP_001139306.1 | 6 - 7 | 1349 | 64 | |
NM_001145835.1 | NP_001139307.1 | 6 - 7 | 1261 | 64 | ||
NM_001145836.1 | NP_001139308.1 | 6 - 7 | 1225 | 64 | ||
NM_009058.2 | NP_033084.2 | 6 - 7 | 1385 | 64 | ||
GenBank mRNA | AK129328.1 | - | 6 - 7 | 1226 | 64 | |
AK141959.1 | - | 6 - 7 | 1216 | 64 | ||
AK149644.1 | - | 1 - 2 | 120 | 64 | ||
AK159020.1 | - | 6 - 7 | 1218 | 64 | ||
AK159754.1 | - | 6 - 7 | 1231 | 64 | ||
AK186985.1 | - | 1 - 2 | 164 | 64 | ||
AK194776.1 | - | 1 - 2 | 164 | 64 | ||
BC027225.1 | - | 6 - 7 | 1043 | 3486732 | 64 | |
L07924.1 | - | 6 - 7 | 1221 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |