Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10465835 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10465836 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10465837 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10465834 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1448996_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10465833 |
- |
- |
Mouse Genome 430 2.0 Array |
1448996_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1448996_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
19881_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
19881_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR1900010081.mm.1 |
NM_009073.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010081.mm.1 |
NM_009073.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900010081.mm.1 |
TC1900001077.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1448996_PM_at |
- |
- |
Mouse Expression Set 430 |
1448996_at |
- |
- |
Murine Genome U74v2 Set |
93453_PM_at |
- |
- |
Murine Genome U74v2 Set |
161525_PM_f_at |
- |
- |
Murine Genome U74v2 Set |
93453_at |
- |
- |
Murine Genome U74v2 Set |
161525_f_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1448996_at |
- |
- |
Mouse Genome 430A 2.0 Array |
1448996_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1448996_PM_at |
- |
- |
Affymetrix HT Mouse Genome 430A Array |
1448996_at |
- |
- |