Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009199.2 | NP_033225.1 | 6 - 7 | 646 | 75 | |
GenBank mRNA | AK018188.1 | - | 6 - 7 | 679 | 75 | |
AK045632.1 | - | 6 - 7 | 678 | 75 | ||
BC064797.1 | - | 6 - 7 | 668 | 30029501 | 75 | |
BC065099.1 | - | 6 - 7 | 670 | 30364069 | 75 | |
D43797.1 | - | 6 - 7 | 652 | 75 | ||
U73521.1 | - | 6 - 7 | 646 | 75 | ||
U75214.1 | - | 2 - 3 | 421 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |