Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009204.2 | NP_033230.2 | 4 - 5 | 657 | 77 | |
GenBank mRNA | AB008453.1 | - | 4 - 5 | 455 | 77 | |
AK137607.1 | - | 4 - 5 | 642 | 77 | ||
AK138832.1 | - | 4 - 5 | 631 | 77 | ||
AK164310.1 | - | 4 - 5 | 632 | 77 | ||
BC014282.1 | - | 4 - 5 | 627 | 4207674 | 77 | |
M23383.1 | - | 4 - 5 | 607 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |