Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253892.1 | NP_001240821.1 | 9 - 10 | 1508 | 98 | |
NM_009207.3 | NP_033233.2 | 10 - 11 | 1704 | 98 | ||
GenBank mRNA | AK145930.1 | - | 10 - 11 | 1629 | 98 | |
AK147858.1 | - | 10 - 11 | 1660 | 98 | ||
BC054102.1 | - | 9 - 10 | 1513 | 30017310 | 98 | |
J04036.1 | - | 10 - 11 | 1619 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |