Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009285.3 | NP_033311.3 | 1 - 2 | 218 | 83 | |
GenBank mRNA | AF099098.1 | - | 1 - 2 | 270 | 83 | |
AK134260.1 | - | 1 - 2 | 219 | 83 | ||
AK142220.1 | - | 1 - 2 | 117 | 83 | ||
AK149293.1 | - | 1 - 2 | 101 | 83 | ||
AK149351.1 | - | 1 - 2 | 101 | 83 | ||
AK161960.1 | - | 1 - 2 | 100 | 83 | ||
BC021425.1 | - | 1 - 2 | 239 | 5010981 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |