Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009309.2 | NP_033335.1 | 3 - 4 | 708 | 87 | |
GenBank mRNA | BC120807.1 | - | 3 - 4 | 680 | 40129730 | 87 |
X51683.1 | - | 3 - 4 | 708 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |