Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009309.2 | NP_033335.1 | 4 - 5 | 782 | 69 | |
GenBank mRNA | BC120807.1 | - | 4 - 5 | 754 | 40129730 | 69 |
X51683.1 | - | 4 - 5 | 782 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |