Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc6a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009320.4 | NP_033346.2 | 6 - 7 | 1025 | 62 | |
GenBank mRNA | AK143510.1 | - | 6 - 7 | 989 | 62 | |
BC015245.1 | - | 6 - 7 | 962 | 4242193 | 62 | |
L03292.2 | - | 6 - 7 | 997 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |