Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbxa2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277265.1 | NP_001264194.1 | 1 - 2 | 217 | 71 | |
GenBank mRNA | AK158477.1 | - | 1 - 2 | 190 | 71 | |
D10849.1 | - | 1 - 2 | 96 | 71 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mus musculus 1700 Inventoried Probe spans exons |