Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009374.3 | NP_033400.2 | 2 - 3 | 245 | 79 | |
GenBank mRNA | BC129890.1 | - | 2 - 3 | 211 | 40111487 | 79 |
BC129891.1 | - | 2 - 3 | 211 | 40111493 | 79 | |
L10385.1 | - | 2 - 3 | 236 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |